missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLXND1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115773
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Plexin D1.
Cell surface receptor for SEMA4A and for class 3 semaphorins, such as SEMA3A, SEMA3C and SEMA3E. Plays an important role in cell-cell signaling, and in regulating the migration of a wide spectrum of cell types. Regulates the migration of thymocytes in the medulla. Regulates endothelial cell migration. Plays an important role in ensuring the specificity of synapse formation. Required for normal development of the heart and vasculature. Mediates anti-angiogenic signaling in response to SEMA3E. [UniProt].
Specifications
| PLXND1 | |
| Polyclonal | |
| Unconjugated | |
| PLXND1 | |
| 6230425C21Rik; b2b1863Clo; b2b553Clo; KIAA0620; MGC75353; PLEXD1; plexin D1; plexin-D1; Plxnd1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23129 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9Y4D7 | |
| PLXND1 | |
| A synthesized peptide derived from human PLXND1(Accession Q9Y4D7), corresponding to amino acid residues W722-L772. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction