missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLXND1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143749
This item is not returnable.
View return policy
Description
The antibody detects 250, 215, and 195 kDa bands corresponding to the apparent molecular mass of Plexin D1 variants in human umbilical vein endothelial cells.
Cell surface receptor for SEMA4A and for class 3 semaphorins, such as SEMA3A, SEMA3C and SEMA3E. Plays an important role in cell-cell signaling, and in regulating the migration of a wide spectrum of cell types. Regulates the migration of thymocytes in the medulla. Regulates endothelial cell migration. Plays an important role in ensuring the specificity of synapse formation. Required for normal development of the heart and vasculature. Mediates anti-angiogenic signaling in response to SEMA3E. [UniProt].
Specifications
| PLXND1 | |
| Polyclonal | |
| Unconjugated | |
| PLXND1 | |
| 6230425C21Rik; b2b1863Clo; b2b553Clo; KIAA0620; MGC75353; PLEXD1; plexin D1; plexin-D1; Plxnd1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23129, 312652, 67784 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| Q3UH93, Q9Y4D7 | |
| PLXND1 | |
| Plexin D1 synthetic peptide (coupled to carrier protein) corresponds to amino acids in the semaphorin domain from human Plexin D1. This sequence is well conserved in rat and mouse Plexin D1, and has low homology to other Plexin family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction