missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLZF Antibody (816421) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB29441JF525
This item is not returnable.
View return policy
Description
PLZF Monoclonal antibody specifically detects PLZF in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
| PLZF | |
| Monoclonal | |
| Janelia Fluor 525 | |
| PLZFzinc finger and BTB domain-containing protein 16, zinc finger and BTB domain containing 16, Zinc finger protein 145, zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia), Zinc finger protein PLZF, ZNF145Promyelocytic leukemia zinc finger protein | |
| E. coli-derived recombinant mouse PLZF, Met1-Gln254, Accession # NP_001028496 | |
| 0.1 mL | |
| Cancer, Innate Immunity, Stem Cell Markers | |
| 7704 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry (Intracellular Staining) | |
| 816421 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction