missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMEL17/SILV Antibody (SPM286), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34755APC
This item is not returnable.
View return policy
Description
PMEL17/SILV Monoclonal specifically detects PMEL17/SILV in Human, Equine, Rat (Negative) samples. It is validated for Flow Cytometry.
Specifications
| PMEL17/SILV | |
| Monoclonal | |
| APC | |
| D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6490 | |
| Human, Equine, Rat (Negative) | |
| Purified |
| Flow Cytometry | |
| SPM286 | |
| Flow Cytometry | |
| PMEL | |
| Membranes from a human melanoma metastasis | |
| 0.1 mL | |
| Primary | |
| By immunohistochemistry, it specifically recognizes a protein in melanocytes and melanomas. This monoclonal antibody reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin. This Mab labels formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction