missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMF1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | PMF1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PMF1 Polyclonal antibody specifically detects PMF1 in Human samples. It is validated for Western BlotSpecifications
| PMF1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 11243 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| PMF-1, Polyamine Modulated Factor 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-205 of human PMF1 (NP_009152.2). MAEASSANLGSGCEEKRHEGSSSESVPPGTTISRVKLLDTMVDTFLQKLVAAGSYQRFTDCYKCFYQLQPAMTQQIYDKFIAQLQTSIREEISDIKEEGNLEAVLNALDKIVEEGKVRKEPAWRPSGIPEKDLHSVMAPYFLQQRDTLRRHVQKQEAENQQLADAVLAGRRQVEELQLQVQAQQQAWQALHREQRELVAVLREPE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title