missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PMFBP1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566035

Product Code. 15918764

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LYRDKYQSSLS NIELLECQVK MLQGELGGIM GQEPENKGDH SKVRIYTSPC MIQEHQETQK RLSEVWQKVS QQDDLIQELR NKLACSNALV LEREKALIKL QADFASCTAT HRYPPSSSEE CEDIKKILKH LQEQKD Highest antigen sequence identity to the following orthologs - mouse 87%, rat 86%.

May play a role in sperm morphology especially the sperm tail and consequently affect fertility. May also be involved in the general organization of cellular cytoskeleton.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PMFBP1
Polyclonal
Unconjugated
PMFBP1
1700016D22Rik; F77; LOW QUALITY PROTEIN: polyamine-modulated factor 1-binding protein 1; ODF3; outer dense fiber ODF3; outer dense fiber protein 3; PMF-1 binding protein; PMF-1-binding protein; PMFBP1; polyamine modulated factor 1 binding protein 1; polyamine-modulated factor 1-binding protein 1; sperm tail associated protein; sperm tail-associated protein; Stap
Rabbit
Antigen Affinity Chromatography
RUO
83449
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8TBY8
PMFBP1
Recombinant Human PMFBP1. Recombinant protein control fragment (Product #RP-106723).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.