Learn More
Kartell™ PMMA (Plexiglas) Two-Tier Test Tube Racks
Kartell™ PMMA (Plexiglas) Two-Tier Test Tube Racks feature smaller holes in the lower tier of the rack to offer stability while safely holding test tubes.
Brand:  Kartell™ 0089100
Additional Details : Weight : 0.31500kg
 View more versions of this product
Product Code. 12001173
  • € 35.25 / Each
In stock, ready to ship: 5
Add to basket
Description
Description
  • Sturdy
  • Made of glass clear PMMA (Plexiglas)
  • Smaller holes in the lower tier of the rack hold the test tubes safely.
Specifications
Specifications
12
40 mm
81 mm
316.5 mm
1 Pack
Two-Tier Test Tube Rack
Clear
Smaller holes in the lower tier of the rack hold the test tubes safely
Tubes (40 mm)
PMMA
12 x 40 mm holes