Learn More
VITLAB™ PMP Urbanti Funnels
Brand:  VITLAB™ 325195
Additional Details : Weight : 0.10800kg
 View more versions of this product
Product Code. 15222159
  • € 110.00 / Pack of 6
Estimated Shipment: 27-05-2024
to see stock.
Specifications
80 mL
PMP
150mm
3mm
210 mm