Learn More
VITLAB™ PMP Urbanti Funnels
Brand:  VITLAB™ 325495
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15252159
  • € 115.00 / Pack of 2
Estimated Shipment: 05-06-2024
to see stock.
Specifications
1800 mL
PMP
155mm
20mm
315 mm