missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PMP22 Monoclonal Antibody (Hu1)

Product Code. 11579892
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11579892 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11579892 Supplier Invitrogen™ Supplier No. MA146002

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Suggested positive control: fetal spinal cord.

Peripheral Myelin Protein 22 (PMP22) is a 160 amino acid glycoprotein which is a member of the claudin family and is thought to play an important role in the structure of peripheral nerve myelin. Irregularities in its gene results in hereditary demyelinating neuropathies. PMP22 protein is present in the compact myelin of adult human peripheral nerve and its distribution mimics that of Myelin Protein zero (P0).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMP22
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone Hu1
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% sodium azide
Gene PMP22
Gene Accession No. Q01453
Gene Alias 22kDa; Cd25; CMT1A; CMT1E; DSS; GAS3; GAS-3; Growth arrest-specific protein 3; HMSNIA; HNPP; peripheral myelin; peripheral myelin protein 22; peripheral myelin protein 22 kDa; PMP22; PMP-22; Protein CD25; SAG; schwann cell membrane glycoprotein; Sp110; SR13 myelin protein; Tr; trembler
Gene Symbols PMP22
Host Species Mouse
Immunogen Human PMP22 cDNA boosted with a 13-amino acid peptide of the 2nd extracellular domain (a.a. 120-133) conjugated to murine IgG. [Swiss-Prot# Q01453].
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5376
Target Species Human, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.