missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMP70 Recombinant Rabbit Monoclonal Antibody (23GB5010), Invitrogen™

Description
Peroxisomes are single membrane organelles which are involved with many important biochemical pathways and are found in almost all eukaryotic cells. Various toxins including, phenols, alcohols, and aldehydes are targeted for modification by peroxisomes. Long chain fatty acid metabolism via beta-oxidation is also mediated by this organelle. Loss of peroxisomal functions may result in diseases such as Zellweger syndrome, hyperpipecolic acidemia, neonatal adrenoleukodystrophy, and infantile Refsum' disease. Peroxisomal membrane protein 70 (PMP70) is an abundant, integral membrane protein of the peroxisome. This protein is induced by treatment with hypolipidemic agents in parallel with peroxisome proliferation and stimulation of the peroxisomal beta-oxidation enzymes.
Specifications
Specifications
| Antigen | PMP70 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB5010 |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ABCD3 |
| Gene Accession No. | P16970, P28288, P55096 |
| Gene Alias | 68 kDa peroxisomal membrane protein; 70 kDa peroxisomal membrane protein; 70-kDa peroxisomal membrane protein; ABC43; ABCD3; AI313901; ATP binding cassette subfamily D member 3; ATP-binding cassette sub-family D member 3; ATP-binding cassette, subfamily D (ALD), member 3; ATP-binding cassette, sub-family D (ALD), member 3; AU018866; AW146054; CBAS5; dJ824O18.1 (ATP-binding cassette, sub-family D (ALD), member 3 (PMP70, PXMP1)); Peroxisomal membrane protein 1; peroxisomal membrane protein 1 (70kD, Zellweger syndrome); peroxisomal membrane protein, 70 kDa; Peroxisomal membrane protein-1 (70kD); PMP68; Pmp70; Pxmp1; ZWS2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?