missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PMPCA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29953
This item is not returnable.
View return policy
Description
PMPCA Polyclonal antibody specifically detects PMPCA in Human samples. It is validated for Immunoprecipitation
Specifications
| PMPCA | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| Alpha-MPPMGC104197, EC 3.4.24.64, inositol polyphosphate-5-phosphatase, 72 kD, INPP5E, KIAA0123FLJ26258, mitochondrial matrix processing protease, alpha subunit, mitochondrial-processing peptidase subunit alpha, MPPA, P-55, peptidase (mitochondrial processing) alpha | |
| Between 475 and 525 | |
| 100 μg | |
| Cell Biology, Endocrinology, Signal Transduction | |
| 23203 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction