Learn More
Invitrogen™ PMS2 Monoclonal Antibody (8E4G11)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA548575
Product Code. 17926311
  • € 498.00 / 100µg
Description
PMS2 Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA
PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PMS2
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P54278
PMS2
Purified recombinant fragment of human PMS2 (AA: 748-851) expressed in E. Coli.
100 μg
Primary
Human
Antibody
IgG2b
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot
8E4G11
Unconjugated
PMS2
AW555130; DNA mismatch repair protein PMS2; H_DJ0042M02.9; HNPCC4; mismatch repair endonuclease PMS2; MLH4; PMS1 homolog 2, mismatch repair protein; PMS1 homolog 2, mismatch repair system component; PMS1 protein homolog 2; PMS2; PMS2 postmeiotic segregation increased 2; PMS2 postmeiotic segregation increased 2 (S. cerevisiae); PMS2CL; PMSL2; postmeiotic segregation increased 2; postmeiotic segregation increased 2 (S. cerevisiae); postmeiotic segregation increased 2 nirs variant 6
Mouse
Protein G
RUO
5395
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.