missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMVK/phosphomevalonate kinase Antibody (21C10), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59412F
This item is not returnable.
View return policy
Description
PMVK/phosphomevalonate kinase Monoclonal antibody specifically detects PMVK/phosphomevalonate kinase in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| PMVK/phosphomevalonate kinase | |
| Monoclonal | |
| FITC | |
| hPMK, HUMPMKI, phosphomevalonate kinase, PMKA, PMKase, PMKEC 2.7.4.2, PMKI | |
| Recombinant human PMVK/phosphomevalonate kinase (1-192aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, metabolism | |
| 10654 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 21C10 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction