missing translation for 'onlineSavingsMsg'
Learn More

PNK Antibody (SR2020), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 349.00 - € 449.00

Specifications

Antigen PNK
Clone SR2020
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18669144
View Documents
Novus Biologicals
NBP3-22567-100ul
100 μg
€ 449.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18656084
View Documents
Novus Biologicals
NBP3-22567-50ul
50 μg
€ 349.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PNK Monoclonal antibody specifically detects PNK in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

PNK
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human
DNA 5'-kinase/3'-phosphatase, EIEE10, Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP), MCSZ, PNKbifunctional polynucleotide phosphatase/kinase, polynucleotide kinase 3'-phosphatase, Polynucleotide kinase-3'-phosphatase
A synthesized peptide derived from human PNK (Uniprot #: Q96T60)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2020
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
DNA Repair, Protein Phosphatase, Signal Transduction
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
11284
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.