missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNK Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Anti-human Polynucleotide Kinase Mouse Monoclonal Antibody recognizes polynucleotide kinase (PNK), a 57 kDa nuclear enzyme implicated in DNA strand repair processes that occur during DNA replication and recombination, or in response to DNA damaging agents. PNK targets the DNA 5'-hydroxy and 3'-phosphate termini for phosphorylation and dephosphorylation events. During strand breakage in human cells, normal PNK associates with other nuclear proteins to prepare the DNA termini for the enzymatic actions of DNA polymerase-beta and DNA ligase III. PNK is a key functional component in maintaining the genetic stability of human cells.
Specifications
Specifications
| Antigen | PNK |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Pnkp |
| Gene Accession No. | Q96T60 |
| Gene Alias | 1810009G08Rik; 2'(3')-polynucleotidase; AOA4; bifunctional polynucleotide phosphatase/kinase; DNA 5'-kinase/3'-phosphatase; EIEE10; Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP); MCSZ; phosphatase/kinase; PKNP; PNK; PNKP; Polynucleotide 3'-phosphatase; Polynucleotide 5'-hydroxyl-kinase; polynucleotide kinase 3'- phosphatase; polynucleotide kinase 3'-phosphatase; polynucleotide kinase-3'-phosphatase |
| Gene Symbols | Pnkp |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?