missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNKD Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98628V
This item is not returnable.
View return policy
Description
PNKD Polyclonal antibody specifically detects PNKD in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PNKD | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| BRP17FKSG19, DKFZp564N1362, DYT8paroxysmal nonkinesiogenic dyskinesia, FPD1brain protein 17, KIAA1184Myofibrillogenesis regulator 1, KIPP1184probable hydrolase PNKD, MGC31943, MR1Paroxysmal nonkinesiogenic dyskinesia protein, MR-1Trans-activated by hepatitis C virus core protein 2, paroxysmal nonkinesigenic dyskinesia, PDCEC 3.-, TAHCCP2PKND1 | |
| Produced in rabbits immunized with E. coli-derived Human PNKD fragment. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Signal Transduction | |
| 25953 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction