missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNLIP Recombinant Rabbit Monoclonal Antibody (HL1310)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA546996
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
This gene is a member of the lipase gene family. It encodes a carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides, and is essential for the efficient digestion of dietary fats. This gene is expressed specifically in the pancreas.
Specifications
| PNLIP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P16233, P27657, Q6P8U6 | |
| PNLIP | |
| Recombinant protein encompassing a sequence within the center region of human Pancreatic Lipase. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL1310 | |
| Unconjugated | |
| PNLIP | |
| 1810007A24Rik; pancreatic lipase; pancreatic triacylglycerol lipase; pancreatic triglyceride lipase; PANLI; PL; PNLIP; PNLIPD; PNLIPRP2; PTL; RP11-33D13.1; triglyceride lipase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 25702, 5406, 69060 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction