Learn More
Invitrogen™ PNMA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553266
Description
Immunogen sequence: RLMESLRGPA ADVIRILKSN NPAITTAECL KALEQVFGSV ESSRDAQIKF LNTYQNPGEK LSAYVIRLEP LLQKVVEKGA IDKDNVNQAR LEQVIAGANH SGAIRRQLWL TGAGEGPAPN LFQLLVQIRE EEAKEEEEEA EATLLQLGL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.
PNMA1 encodes a protein that is highly restricted to the brain and testis. Anti-PNMA1 reacts mainly with subnuclear elements (including the nucleoli) and to a lesser degree the cytoplasm.
Specifications
| PNMA1 | |
| Polyclonal | |
| Unconjugated | |
| Pnma1 | |
| 37 kDa neuronal protein; 5730402C15Rik; MA1; Neuron- and testis-specific protein 1; paraneoplastic antigen MA1; Paraneoplastic antigen Ma1 homolog; paraneoplastic Ma antigen 1; paraneoplastic neuronal antigen MA1; paraneoplastic onconeuronal protein MA1; PNMA family member 1; PNMA1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9240 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8ND90 | |
| Pnma1 | |
| Recombinant protein corresponding to Human PNMA1. Recombinant protein control fragment (Product #RP-88885). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.