missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMA3 Antibody (OTI2E6), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73487AF594
This item is not returnable.
View return policy
Description
PNMA3 Monoclonal specifically detects PNMA3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PNMA3 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PNMA3 | |
| Recombinant protein fragment corresponding to amino acids 1-250 of human PNMA3 (NP_037496) produced in E.coli. | |
| 0.1 mL | |
| 29944.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2E6 | |
| Western Blot, Immunohistochemistry | |
| MA3MA5, MGC132756, MGC132758, paraneoplastic antigen MA3, paraneoplastic cancer-testis-brain antigen | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction