missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMA3 Antibody (OTI2E6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73487
This item is not returnable.
View return policy
Description
PNMA3 Monoclonal specifically detects PNMA3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PNMA3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500, Immunohistochemistry 1:150 | |
| MA3MA5, MGC132756, MGC132758, paraneoplastic antigen MA3, paraneoplastic cancer-testis-brain antigen | |
| Mouse | |
| 52.2 kDa | |
| 100 μg | |
| 29944 | |
| Human | |
| Lyophilized |
| Western Blot, Immunohistochemistry | |
| OTI2E6 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| PNMA3 | |
| Human recombinant corresponding to amino acids 1-250 of human PNMA3(NP_037496) produced in E.coli. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction