missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMA3 Antibody (OTI2E6), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73487R
This item is not returnable.
View return policy
Description
PNMA3 Monoclonal specifically detects PNMA3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PNMA3 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PNMA3 | |
| Recombinant protein fragment corresponding to amino acids 1-250 of human PNMA3 (NP_037496) produced in E.coli. | |
| 0.1 mL | |
| 29944.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2E6 | |
| Western Blot, Immunohistochemistry | |
| MA3MA5, MGC132756, MGC132758, paraneoplastic antigen MA3, paraneoplastic cancer-testis-brain antigen | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?