missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMA3 Mouse anti-Human, Clone: OTI1C3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502336
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the paraneoplastic antigen MA (PNMA) family, which shares homology with retroviral Gag proteins. The PNMA antigens are highly expressed in the brain and also in a range of tumors associated with serious neurological phenotypes. There are reports of the presence of a functional -1 ribosomal frameshift signal (consisting of a heptanucleotide shift motif followed 3′ by a pseudoknot structure) in this gene, however, the frame-shifted product has not been characterized. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| PNMA3 | |
| Monoclonal | |
| 0.65 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UL41 | |
| PNMA3 | |
| Full length human recombit protein of human PNMA3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C3 | |
| Unconjugated | |
| PNMA3 | |
| DmelCG13488 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29944 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction