missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMA3 Mouse anti-Human, Clone: OTI2D3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502466
This item is not returnable.
View return policy
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the paraneoplastic antigen MA (PNMA) family, which shares homology with retroviral Gag proteins. The PNMA antigens are highly expressed in the brain and also in a range of tumors associated with serious neurological phenotypes. There are reports of the presence of a functional -1 ribosomal frameshift signal (consisting of a heptanucleotide shift motif followed 3′ by a pseudoknot structure) in this gene, however, the frame-shifted product has not been characterized. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| PNMA3 | |
| Monoclonal | |
| Unconjugated | |
| PNMA3 | |
| DmelCG13488 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29944 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2D3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UL41 | |
| PNMA3 | |
| Full length human recombit protein of human PNMA3 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?