missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNMT Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1D2, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502822S
This item is not returnable.
View return policy
Description
The product of this gene catalyzes the last step of the catecholamine biosynthesis pathway, which methylates norepinephrine to form epinephrine (adrenaline). The enzyme also has beta-carboline 2N-methyltransferase activity. This gene is thought to play a key step in regulating epinephrine production.Specifications
| PNMT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10937, P11086, P40935 | |
| PNMT | |
| Full length human recombit protein of human PNMT produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D2 | |
| Unconjugated | |
| PNMT | |
| PENT, PNMTase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 18948, 24661, 491023, 5409 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction