Learn More
PNMT Mouse anti-Human, Clone: OTI1B7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502825
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The product of this gene catalyzes the last step of the catecholamine biosynthesis pathway, which methylates norepinephrine to form epinephrine (adrenaline). The enzyme also has beta-carboline 2N-methyltransferase activity. This gene is thought to play a key step in regulating epinephrine production.Specifications
| PNMT | |
| Monoclonal | |
| Unconjugated | |
| PNMT | |
| PENT, PNMTase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5409 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1B7 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P11086 | |
| PNMT | |
| Full length human recombit protein of human PNMT produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.