missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PNO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570579
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
Positively regulates dimethylation of two adjacent adenosines in the loop of a conserved hairpin near the 3'-end of 18S rRNA (PubMed:25851604). [UniProt].
Specifications
| PNO1 | |
| Polyclonal | |
| Unconjugated | |
| PNO1 | |
| 1810003N24Rik; AA407421; AU015468; AU019495; AU042323; Emi3; expressed during mesenchymal induction 3; Imi3; induced during mesenchymal induction 3; KHRBP1; KH-type RNA-binding protein 1; partner of NOB1 homolog; partner of NOB1 homolog (S. cerevisiae); Pno1; putatative 28 kDa protein; putative 28 kDa protein; RNA binding protein; RNA-binding protein PNO1; RRP20 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56902 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NRX1 | |
| PNO1 | |
| synthetic peptide directed towards the middle region of human PNO1 (aa 31-80). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction