missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPO Antibody (2C7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97941F
This item is not returnable.
View return policy
Description
PNPO Monoclonal specifically detects PNPO in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PNPO | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| PNPO | |
| Recombinant human PNPO (57-261aa) purified from E. coli | |
| 100 μL | |
| Cancer, metabolism | |
| 55163 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2C7 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| EC 1.4.3.5, FLJ10535, PDXPO, pyridoxal 5'-phosphate synthase, pyridoxamine 5'-phosphate oxidase, Pyridoxamine-phosphate oxidase, pyridoxine 5'-phosphate oxidase, pyridoxine-5'-phosphate oxidase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human PNPO mAb, clone 2C7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PNPO protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction