missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPO Antibody (2C7), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97941MFV450
This item is not returnable.
View return policy
Description
PNPO Monoclonal antibody specifically detects PNPO in Human samples. It is validated for Flow Cytometry
Specifications
| PNPO | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2C7 | |
| Flow Cytometry | |
| EC 1.4.3.5, FLJ10535, PDXPO, pyridoxal 5'-phosphate synthase, pyridoxamine 5'-phosphate oxidase, Pyridoxamine-phosphate oxidase, pyridoxine 5'-phosphate oxidase, pyridoxine-5'-phosphate oxidase | |
| Recombinant human PNPO (57-261aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, metabolism | |
| 55163 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction