missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPO Antibody (OTI1H9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73508CL1
This item is not returnable.
View return policy
Description
PNPO Monoclonal antibody specifically detects PNPO in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| PNPO | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 1.4.3.5, FLJ10535, PDXPO, pyridoxal 5'-phosphate synthase, pyridoxamine 5'-phosphate oxidase, Pyridoxamine-phosphate oxidase, pyridoxine 5'-phosphate oxidase, pyridoxine-5'-phosphate oxidase | |
| Full length human recombinant protein of human PNPO(NP_060599) produced in HEK293T cell. | |
| 0.1 mL | |
| Cancer, metabolism | |
| 55163 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| OTI1H9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction