missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPO Monoclonal Antibody (OTI2F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503178S
This item is not returnable.
View return policy
Description
PNPO Monoclonal antibody specifically detects PNPO in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| PNPO | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96EK2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 112885 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI2F8 | |
| Unconjugated | |
| PHF21B | |
| AI415282; epididymis secretory protein Li 302; FLJ10535; HEL-S-302; PDXPO; Pnpo; pyridoxal 5'-phosphate synthase; pyridoxamine 5'-phosphate oxidase; pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; pyridoxine 5-phosphate oxidase; pyridoxine-5'-phosphate oxidase | |
| Full length human recombinant protein of human PHF21B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction