missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNR/NR2E3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS5770
This item is not returnable.
View return policy
Description
PNR/NR2E3 Polyclonal antibody specifically detects PNR/NR2E3 in Human, Mouse, Porcine, Bat, Bovine, Equine, Hamster samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PNR/NR2E3 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Nuclear receptor subfamily 2 group E member 3, nuclear receptor subfamily 2, group E, member 3, photoreceptor-specific nuclear receptor, PNRESCS, rd7, Retina-specific nuclear receptor, RNR, RP37MGC49976 | |
| Synthetic 19 amino acid peptide from ligand-binding domain of human PNR/NR2E3. | |
| 0.05 mL | |
| Vision | |
| 10002 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 30 μg/mL | |
| Q9Y5X4 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Bat, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction