missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PNU 120596

Description
PNU 120596 is a positive allosteric modulator of α7 neuronal nicotinic acetylcholine receptors (EC50 = 216 nM), with no detectable effect on α4β2, α3β4 and α9α10 receptors. Active in vivo following systemic administration. Neuroprotective in an in vivo model of transient focal cerebral ischemia.
Specifications
Specifications
| Chemical Name or Material | N-(5-Chloro-2,4-dimethoxyphenyl)-N'-(5-methyl-3-isoxazolyl)-urea |
| CAS | 501925-31-1 |
| Assay Percent Range | >99% |
| Quantity | 5 mg |
| Molecular Formula | C13H14ClN3O4 |
| Formula Weight | Observed MW: 311.72 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?