missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POC5 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78741PE
This item is not returnable.
View return policy
Description
POC5 Polyclonal antibody specifically detects POC5 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| POC5 | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| centrosomal protein POC5, FLJ35779, POC5 centriolar protein homolog (Chlamydomonas) | |
| The immunogen for this product maps to a region between residue 525 and 575 of human Protein of Centriole 5 using the numbering given in entry NP_001092741.1 (GeneID 134359). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 134359 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction