missing translation for 'onlineSavingsMsg'
Learn More

PODNL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 220.00 - € 307.65

Specifications

Antigen PODNL1
Dilution Immunohistochemistry-Paraffin 1:50-1:200
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18699796
View Documents
Novus Biologicals
NBP3-05990-100ul
100 μg
ONLINE EXCLUSIVE PRICE
€ 326.00
€ 307.65 / 100µL
Save € 18.35
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18643187
View Documents
Novus Biologicals
NBP3-05990-50ul
50 μg
€ 220.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PODNL1 Polyclonal antibody specifically detects PODNL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

PODNL1
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
FLJ23447, podocan-like 1, podocan-like protein 1, SLRR5B
Produced in rabbits immunized with E. coli-derived Human PODNL1 fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Purified
RUO
PBS, pH7.0
79883
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.