missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podocalyxin Like Antibody (192703) [DyLight 755], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1556Z
This item is not returnable.
View return policy
Description
Podocalyxin Like Monoclonal antibody specifically detects Podocalyxin Like in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Specifications
| Podocalyxin Like | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 192703 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready | |
| Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Podocalyxin, Ser21-Arg402, Accession # Q9R0M4 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 5420 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction