Learn More
Podocalyxin Like Antibody (192703), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB1556MFV450
Product Code. 18564455
  • € 694.00 / 0.10mL
Description
Podocalyxin Like Monoclonal antibody specifically detects Podocalyxin Like in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Podocalyxin Like
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Rat
0.1 mL
Cancer, Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers
5420
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
192703
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1
Mouse myeloma cell line NS0-derived recombinant mouse Podocalyxin, Ser21-Arg402, Accession # Q9R0M4
RUO
Primary
Mouse
IgG2b