missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Podocin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561987
This item is not returnable.
View return policy
Description
Immunogen sequence: KAASESLRMA AEILSGTPAA VQLRYLHTLQ SLSTEKPSTV VLPLPFDLLN CLSSPSNRTQ GSLPFPSPSK PVEPLNPK Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
The pathological conditions of SRN1 correlate well with mutations at the NPHS2 gene, where expression of a protein known as Podocin occurs. Abnormal or inefficient signaling through Podocin protein-dependent networks contributes to the development of podocyte dysfunction and proteinuria. The human NPHS2 gene maps to chromosome 1q25-q31 and encodes a 383 amino acid protein. Podocin is an integral membrane protein that appears to fold into a hairpin-like structure with intracellular amino- and carboxy-termini. Transmembrane and cytoplasmic portions of Podocin share homology to the corresponding regions of the stomatin family proteins. Expression of high-order oligomers of Podocin in glomerular podocytes may reflect a scaffolding function that influences proper function of the glomerular filtration barrier, which is necessary for renal stability.
Specifications
| Podocin | |
| Polyclonal | |
| Unconjugated | |
| Nphs2 | |
| AI790225; nephrosis 2 homolog, podocin; nephrosis 2 idiopathic steroid-resistant (podocin); nephrosis 2, idiopathic, steroid-resistant; nephrosis 2, idiopathic, steroid-resistant (podocin); nephrosis 2, podocin; NPHS2; NPHS2 podocin; NPHS2 stomatin family member, podocin; NPHS2, podocin; PDCN; podocin; SRN1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7827 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NP85 | |
| Nphs2 | |
| Recombinant protein corresponding to Human Podocin. Recombinant protein control fragment (Product #RP-95614). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto