missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (18H5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1013AF647
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal specifically detects Podoplanin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Podoplanin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Q86YL7 | |
| PDPN | |
| MDCK cells stably expressing gp36 (Podoplanin) protein | |
| 0.1 mL | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 10630 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 18H5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human. Not yet tested in other species. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction