missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (8.1.1) - Azide and BSA Free, Novus Biologicals™
Golden Syrian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP2-80916
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal specifically detects Podoplanin in Mouse, Human (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, CyTOF-ready.
Specifications
| Podoplanin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000-1:2000, Flow Cytometry : 1:400, Immunohistochemistry : 1:100-1:500, Immunocytochemistry/Immunofluorescence : 1-5 μg/mL, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin : 1:100-1:500, Immunohistochemistry-Frozen, Electron Microscopy, CyTOF-ready | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| Golden Syrian Hamster | |
| Protein G purified | |
| RUO | |
| Primary | |
| Does not cross-react with human. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 8.1.1 | |
| Unconjugated | |
| PBS | |
| PDPN | |
| Murine thymic stromal cell lines | |
| 0.1 mL | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 10630 | |
| Mouse, Human (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction