missing translation for 'onlineSavingsMsg'
Learn More

Podoplanin Antibody (PDPN/1433), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-54347AF594

Product Code. 18219112

  • € 602.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Podoplanin Monoclonal specifically detects Podoplanin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Podoplanin
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
PDPN
Recombinant human Podoplanin protein fragment (around aa 24-126) (exact sequence is proprietary) (Uniprot: Q86YL7)
0.1 mL
Primary
Human
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
PDPN/1433
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Mouse
Protein A or G purified
Core ESC Like Genes, Signal Transduction, Stem Cell Markers
10630.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.