missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (PDPN/4009R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08816AF700
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Podoplanin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| Synthetic peptide corresponding to residues within aa1-100 of Podoplanin (Uniprot: Q86YL7) | |
| 100 μg | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 10630 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PDPN/4009R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction