missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (pmab-1), Janelia Fluor™ 549, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-03955JF549
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Podoplanin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDPN | |
| The immunogen is Podoplanin. | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2a κ |
| Western Blot, Flow Cytometry | |
| pmab-1 | |
| Western Blot, Flow Cytometry | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| Rat | |
| Protein G purified | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 10630.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction