missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (rPDPN/6994) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20917JF669
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Podoplanin | |
| Monoclonal | |
| Janelia Fluor 669 | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| Recombinant human Podoplanin protein fragment (around aa 1-100) (exact sequence is proprietary) | |
| 0.1 mL | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 10630 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rPDPN/6994 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction