missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Hamster anti-Mouse, PE, Clone: 8.1.1, BD Pharmingen™
Hamster Monoclonal Antibody
Brand: BD Biosciences 566390
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Podoplanin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Hamster | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 8.1.1 | |
| PE | |
| 0.1 mg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction