missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PODXL Monoclonal Antibody (OTI9C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807762S
This item is not returnable.
View return policy
Description
PODXL Monoclonal antibody specifically detects PODXL in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| PODXL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92913 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2258 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI9C2 | |
| Unconjugated | |
| FGF13 | |
| AW121214; GCTM-2 antigen; Gp200; Ly102; MGC138240; PC; PCLP; PCLP antibody; Pclp1; PCLP-1; PCLP1 antibody; PODLX2; podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; podolyxin; Podxl | |
| Full length human recombinant protein of human FGF13 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction