missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PODXL2 Monoclonal Antibody (OTI10D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810168
This item is not returnable.
View return policy
Description
Podolyxin is a CD34 family member expressed by podocytes, vascular endothelium, mesothelium, and a subset of hematopoietic progenitors. It is the major sialoprotein in the glycocalyx of glomerular podocytes. The physiologic role of podolyxin in leukemic blasts is unknown. However, it is thought of as a candidate prostate cancer tumor aggressiveness gene. Podolyxin appears to complement CD34 as a useful cell surface marker for hemangioblasts, the common precursors of hematopoietic and endothelial cells.Specifications
| PODXL2 | |
| Monoclonal | |
| Unconjugated | |
| PODXL2 | |
| EG, PODLX2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 50512 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI10D8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NZ53 | |
| PODXL2 | |
| Human recombit protein fragment corresponding to amino acids 366-486 of human PODXL2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction