missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PODXL2 Monoclonal Antibody (OTI7E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810169S
This item is not returnable.
View return policy
Description
PODXL2 Monoclonal antibody specifically detects PODXL2 in Human samples. It is validated for Western BlotSpecifications
| PODXL2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NZ53 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 50512 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7E1 | |
| Unconjugated | |
| PODXL2 | |
| D130074J02Rik; EG; endoglycan; LOW QUALITY PROTEIN: podocalyxin-like protein 2; PODLX2; podocalyxin like 2; podocalyxin-like 2; podocalyxin-like protein 2; Podxl2; RGD1305815; sialomucin protein; UNQ1861/PRO3742 | |
| Human recombinant protein fragment corresponding to amino acids 366-486 of human PODXL2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction