missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PODXL2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583597

Product Code. 16320555

  • € 572.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EAQSRIPWDS TQVICKDWSN LAGKNYIILN MTENIDCEVF RQHRGPQLLA LVEEVLPRHG SGHHGAWHIS LSKPSEKEQH LLMTLVGEQG VVPTQDVLSM LGDIRRSLEE IGIQNYSTTS S Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 89%.

Podolyxin is a CD34 family member expressed by podocytes, vascular endothelium, mesothelium, and a subset of hematopoietic progenitors. It is the major sialoprotein in the glycocalyx of glomerular podocytes. The physiologic role of podolyxin in leukemic blasts is unknown. However, it is thought of as a candidate prostate cancer tumor aggressiveness gene. Podolyxin appears to complement CD34 as a useful cell surface marker for hemangioblasts, the common precursors of hematopoietic and endothelial cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PODXL2
Polyclonal
Unconjugated
PODXL2
D130074J02Rik; EG; endoglycan; LOW QUALITY PROTEIN: podocalyxin-like protein 2; PODLX2; podocalyxin like 2; podocalyxin-like 2; podocalyxin-like protein 2; Podxl2; RGD1305815; sialomucin protein; UNQ1861/PRO3742
Rabbit
Antigen affinity chromatography
RUO
50512
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NZ53
PODXL2
Recombinant protein corresponding to Human PODXL2. Recombinant protein control fragment (Product #RP-102923).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.