missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POF1B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93152-0.02ml
This item is not returnable.
View return policy
Description
POF1B Polyclonal antibody specifically detects POF1B in Human, Rat samples. It is validated for Western Blot
Specifications
| POF1B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ22792, POF2B, premature ovarian failure, 1B, protein POF1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 330-560 of human POF1B (NP_079197.3). LVLSTFSNIREELGHLQNDMTSLENDKMRLEKDLSFKDTQLKEYEELLASVRANNHQQQQGLQDSSSKCQALEENNLSLRHTLSDMEYRLKELEYCKRNLEQENQNLRMQVSETCTGPMLQAKMDEIGNHYTEMVKNLRMEKDREICRLRSQLNQYHKDVSKREGSCSDFQFKLHELTSLLEEKDSLIKRQSEELSKLRQEIYSSHNQPSTGGRTTITTKKYRTQYPILGL | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 79983 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction